accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3O(E)

CAS RN 2892-18-4

IUPAC Name: (1E)-5-methyl-1-phenylhex-1-en-3-one
Synonyms: (1E)-5-methyl-1-phenylhex-1-en-3-one
Molecular Weight (g/mol): 188.27
Molecular Formula: C13H16O
InChi Key: LLVCDRTZBYXKII-CMDGGOBGSA-N
SMILES: CC(C)CC(=O)\C=C\C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results