Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Styryl Ketone, TCI America™
SDP

Catalog No. I025325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I025325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I025325ML Supplier TCI America Supplier No. I025325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2892-18-4
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
MDL Number MFCD00026487
InChI Key LLVCDRTZBYXKII-CMDGGOBGSA-N
Synonym 5-Methyl-1-phenyl-1-hexen-3-one
PubChem CID 5354491
IUPAC Name (E)-5-methyl-1-phenylhex-1-en-3-one
SMILES CC(C)CC(=O)C=CC1=CC=CC=C1

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 188.27
Physical Form Liquid
Chemical Name or Material Isobutyl Styryl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.