accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2895-21-8

IUPAC Name: 2-chloro-N-(propan-2-yl)acetamide
Synonyms: 2-chloro-N-isopropylacetamide
Molecular Weight (g/mol): 135.59
Molecular Formula: C5H10ClNO
InChi Key: GYPNJSBBOATUPK-UHFFFAOYSA-N
SMILES: CC(C)NC(=O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results