accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OH(S)NH2OHO

CAS RN 28954-12-3

IUPAC Name: (2S,3S)-2-amino-3-hydroxybutanoic acid
Synonyms: L-allothreonine|allo-L-threonine|allothreonine
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: AYFVYJQAPQTCCC-HRFVKAFMSA-N
SMILES: C[C@H](O)[C@H](N)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)