Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L(+)-allo-Threonine, 99%

Catalog No. AC198542500
Change view
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC198542500 250 mg Glass Bottle
AC198541000 100 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC198542500 Supplier Thermo Scientific Chemicals Supplier No. 198542500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 28954-12-3 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28954-12-3
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064268
InChI Key AYFVYJQAPQTCCC-HRFVKAFMSA-N
Synonym l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine
PubChem CID 99289
ChEBI CHEBI:28718
IUPAC Name (2S,3S)-2-amino-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@H](N)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH(NH2)CO2H
Beilstein 04, III, 1629
Specific Rotation +8° to +10° (20°C, 589nm) (c=2, water)
Specific Rotation Condition + 9.00 (20.00°C c=2,H2O)
Quantity 250 mg
Formula Weight 119.12
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material L(+)-allo-Threonine, 99%
Show More Show Less
EINECSNumber 249-327-2
RTECSNumber BA4055000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.