CAS RN 28989-50-6
IUPAC Name:
4-(thiophen-2-yl)-1,3-thiazol-2-amine
Synonyms:
4-(thiophen-2-yl)-1,3-thiazol-2-amine
Molecular Weight (g/mol):
182.26
Molecular Formula:
C7H6N2S2
InChi Key:
XRNPDKQNHIKARG-UHFFFAOYSA-N
SMILES:
NC1=NC(=CS1)C1=CC=CS1
1
–
1
of
1
results
1
2-Amino-4-(2-thienyl)thiazole, 97%, Thermo Scientific™
CAS: 28989-50-6 Molecular Formula: C7H6N2S2 Molecular Weight (g/mol): 182.259 MDL Number: MFCD00457532 InChI Key: XRNPDKQNHIKARG-UHFFFAOYSA-N Synonym: 4-thiophen-2-yl thiazol-2-amine,4-thiophen-2-yl-thiazol-2-ylamine,4-thiophen-2-yl-1,3-thiazol-2-amine,4-2-thienyl-1,3-thiazol-2-amine,2-amino-4-2-thienyl thiazole,2-thiazolamine, 4-2-thienyl,4-2-thienyl-1,3-thiazole-2-ylamine,ncimech_000023,ncistruc1_000175,ncistruc2_000001 PubChem CID: 243973 IUPAC Name: 4-thiophen-2-yl-1,3-thiazol-2-amine SMILES: C1=CSC(=C1)C2=CSC(=N2)N