CAS RN 2899-36-7
CAS RN 2899-36-7
IUPAC Name:
hydrogen ethyl 2-amino-4-(methylsulfanyl)butanoate chloride
Synonyms:
hydrogen ethyl 2-amino-4-(methylsulfanyl)butanoate chloride
Molecular Weight (g/mol):
213.72
Molecular Formula:
C7H16ClNO2S
InChi Key:
KPWCQEUBJAIORR-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CCOC(=O)C(N)CCSC
1
–
1
of
1
results
1
Advanced Chem Tech H-Met-OEt.HCI, Advanced ChemTech
CAS: 2899-36-7 Molecular Formula: C7H16ClNO2S Molecular Weight (g/mol): 213.72 InChI Key: KPWCQEUBJAIORR-UHFFFAOYNA-N IUPAC Name: hydrogen ethyl 2-amino-4-(methylsulfanyl)butanoate chloride SMILES: [H+].[Cl-].CCOC(=O)C(N)CCSC