accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(S)NHO

CAS RN 2901-76-0

IUPAC Name: (2S)-3-phenyl-2-(phenylformamido)propanoate
Synonyms: (2S)-3-phenyl-2-(phenylformamido)propanoate
Molecular Weight (g/mol): 268.29
Molecular Formula: C16H14NO3
InChi Key: NPKISZUVEBESJI-AWEZNQCLSA-M
SMILES: [O-]C(=O)[C@H](CC1=CC=CC=C1)NC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results