Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl-DL-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. B030210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B030210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B030210G Supplier TCI America Supplier No. B030210G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2901-76-0
Molecular Formula C16H15NO3
Molecular Weight (g/mol) 269.3
MDL Number MFCD00037252
InChI Key NPKISZUVEBESJI-UHFFFAOYSA-N
PubChem CID 273362
IUPAC Name 2-benzamido-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)NC(=O)C2=CC=CC=C2

Specifications

Melting Point 187°C
Color White
Quantity 10 g
Formula Weight 269.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoyl-DL-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.