CAS RN 29079-00-3
IUPAC Name:
4-ethynyl-1,1'-biphenyl
Synonyms:
4-ethynyl-1,1'-biphenyl
Molecular Weight (g/mol):
178.23
Molecular Formula:
C14H10
InChi Key:
BPBNKCIVWFCMJY-UHFFFAOYSA-N
SMILES:
C#CC1=CC=C(C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich 4-Ethynylbiphenyl
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.