accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 291-64-5

IUPAC Name: cycloheptane
Synonyms: cycloheptane
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: DMEGYFMYUHOHGS-UHFFFAOYSA-N
SMILES: C1CCCCCC1
Boiling Point 
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)