accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29102-67-8

IUPAC Name: 3,5-dibromo-3',5'-bis(3,5-dibromophenyl)-1,1'-biphenyl
Synonyms: 3,5-dibromo-3',5'-bis(3,5-dibromophenyl)-1,1'-biphenyl
Molecular Weight (g/mol): 779.78
Molecular Formula: C24H12Br6
InChi Key: VBMGLVNBADCRDN-UHFFFAOYSA-N
SMILES: BrC1=CC(=CC(Br)=C1)C1=CC(=CC(=C1)C1=CC(Br)=CC(Br)=C1)C1=CC(Br)=CC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results