Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Tris(3,5-dibromophenyl)benzene 95.0+%, TCI America™
SDP

Catalog No. T3213200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3213200MG 200 mg
T32131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3213200MG Supplier TCI America Supplier No. T3213200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29102-67-8
Molecular Formula C24H12Br6
Molecular Weight (g/mol) 779.78
MDL Number MFCD13193235
InChI Key VBMGLVNBADCRDN-UHFFFAOYSA-N
PubChem CID 591077
IUPAC Name 3,5-dibromo-3',5'-bis(3,5-dibromophenyl)-1,1'-biphenyl
SMILES BrC1=CC(=CC(Br)=C1)C1=CC(=CC(=C1)C1=CC(Br)=CC(Br)=C1)C1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 315°C
Quantity 200 mg
Formula Weight 779.78
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(3,5-dibromophenyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.