accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHNN

CAS RN 29110-74-5

IUPAC Name: 3-chloro-2H-indazole
Synonyms: 3-chloro-2H-indazole
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: QPHAGNNWDZSKJH-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results