accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONONH

CAS RN 29113-33-5

IUPAC Name: ethyl 4-oxo-1,4-dihydroquinazoline-2-carboxylate
Synonyms: ethyl 4-oxo-1H-quinazoline-2-carboxylate
Molecular Weight (g/mol): 218.21
Molecular Formula: C11H10N2O3
InChi Key: BMCAWNQKVVTNFP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NC(=O)C2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results