accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2912-62-1

IUPAC Name: 2-chloro-2-phenylacetyl chloride
Synonyms: 2-chloro-2-phenylacetyl chloride
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: FGEAOSXMQZWHIQ-UHFFFAOYNA-N
SMILES: ClC(C(Cl)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)