Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Chloro-2-phenylacetyl Chloride 97%, Thermo Scientific™

Catalog No. AC157380500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC157380500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC157380500 Supplier Thermo Scientific Chemicals Supplier No. 157380500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2912-62-1
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
InChI Key FGEAOSXMQZWHIQ-UHFFFAOYSA-N
Synonym chloro phenyl acetyl chloride, alpha-chlorophenylacetyl chloride, a-chlorophenylacetyl chloride, .alpha.-chlorophenylacetyl chloride, dl-2-chloro-2-phenylacetyl chloride, ccris 8624, 2-chloro-2-phenylacetylchloride, benzeneacetyl chloride, .alpha.-chloro, chloro phenyl acetylchloride, chlorophenyl acetyl chloride
PubChem CID 102902
IUPAC Name 2-chloro-2-phenylacetyl chloride
SMILES C1=CC=C(C=C1)C(C(=O)Cl)Cl

Specifications

Color Yellow
Infrared Spectrum Authentic
Refractive Index 1.5450 to 1.5470 (20°C, 589nm)
Linear Formula C6H5CH(Cl)COCl
Quantity 50 g
Beilstein 09, 449
Formula Weight 189.04
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material DL-2-Chloro-2-phenylacetyl chloride, 97%
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.