accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrONH2

CAS RN 29124-57-0

IUPAC Name: 2-amino-5-bromobenzaldehyde
Synonyms: 2-amino-5-bromobenzaldehyde
Molecular Weight (g/mol): 200.04
Molecular Formula: C7H6BrNO
InChi Key: VBYZWJMZASVGNB-UHFFFAOYSA-N
SMILES: NC1=C(C=O)C=C(Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)