Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromobenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. A3063200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3063200MG 200 mg
A30631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3063200MG Supplier TCI America Supplier No. A3063200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29124-57-0
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD10696879
InChI Key VBYZWJMZASVGNB-UHFFFAOYSA-N
Synonym 2-amino-5-bromobenzenecarbaldehyde, 5-bromoanthranilaldehyde, 4-bromo-2-formylaniline, benzaldehyde,2-amino-5-bromo, benzaldehyde, 2-amino-5-bromo, pubchem19693, aminobromobenzenecarbaldehyde, 2-amino-5-bromobenzaidehyde, 2-amino-5-bromo-benzaldehyde
PubChem CID 23510475
IUPAC Name 2-amino-5-bromobenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)N

Specifications

Melting Point 78°C
Color Yellow
Quantity 200 mg
Formula Weight 200.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.