accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 292-64-8

IUPAC Name: cyclooctane
Synonyms: cyclooctane
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: WJTCGQSWYFHTAC-UHFFFAOYSA-N
SMILES: C1CCCCCCC1
Boiling Point 
  • (1)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)