accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNOOHOO

CAS RN 2922-40-9

IUPAC Name: 2-amino-3-(4-nitrophenyl)propanoic acid
Synonyms: p-nitrophenylalanine P-nitrophenylalanine 4-nitrophenylalanine
Molecular Weight (g/mol): 210.19
Molecular Formula: C9H10N2O4
InChi Key: GTVVZTAFGPQSPC-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results