Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Chloropurine Riboside

Catalog No. 8524815G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-248-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 85-248-15G Supplier Sigma Aldrich Supplier No. 8524815G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

6-Chloropurine Riboside

Specifications

CAS 2922-40-9
Melting Point 236°C to 237°C
Molecular Formula C9H10N2O4
Linear Formula O2NC6H4CH2CH(NH2)CO2H
MDL Number MFCD00007384
Quantity 5 g
Synonym 6-Chloropurine-9-beta-D-ribofuranoside
Molecular Weight (g/mol) 210.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.