accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29241-60-9

IUPAC Name: 5-bromo-2-chloro-3-methylpyridine
Synonyms: 5-bromo-2-chloro-3-methylpyridine
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: YTSKHJMMNFPBBZ-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CN=C1Cl
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)