Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chloro-3-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. B37445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37445G 5 g
B374425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37445G Supplier TCI America Supplier No. B37445G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29241-60-9
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD03095093
InChI Key YTSKHJMMNFPBBZ-UHFFFAOYSA-N
Synonym 2-chloro-3-methyl-5-bromopyridine, 5-bromo-2-chloro-3-picoline, 5-bromo-2-chloro-3-methyl pyridine, 5-bromo-2-chloro-3-methyl-pyridine, pyridine, 5-bromo-2-chloro-3-methyl, 2-chloro-5-bromo-3-picoline, 2-chloro-3-methyl-5-bromo pyridine, pubchem1104, ksc495o9j
PubChem CID 285434
IUPAC Name 5-bromo-2-chloro-3-methylpyridine
SMILES CC1=CC(=CN=C1Cl)Br

Specifications

Melting Point 44°C
Color White
Boiling Point 78°C
Quantity 5 g
Formula Weight 206.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chloro-3-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.