accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2937-83-9

IUPAC Name: 5-(piperidin-1-yl)pentan-1-ol
Synonyms: 5-(piperidin-1-yl)pentan-1-ol
Molecular Weight (g/mol): 171.28
Molecular Formula: C10H21NO
InChi Key: GEPOCRCIIKQXSM-UHFFFAOYSA-N
SMILES: OCCCCCN1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results