Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Piperidinepentanol 98.0+%, TCI America™
SDP

Catalog No. P13855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13855G 5 g
P13851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13855G Supplier TCI America Supplier No. P13855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2937-83-9
Molecular Formula C10H21NO
Molecular Weight (g/mol) 171.284
MDL Number MFCD03093640
InChI Key GEPOCRCIIKQXSM-UHFFFAOYSA-N
Synonym 1-(5-Hydroxypentyl)piperidine
PubChem CID 566537
IUPAC Name 5-piperidin-1-ylpentan-1-ol
SMILES C1CCN(CC1)CCCCCO

Specifications

Boiling Point 140°C
Quantity 5 g
Formula Weight 171.28
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Piperidinepentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.