accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSOONOOHOHOH3CNOHOOOHOHH2ONa

CAS RN 29412-85-9

IUPAC Name: 2-({[3-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-2,5-dimethylphenyl)-2,2-dioxo-3H-1,2λ⁶-benzoxathiol-3-yl]methyl}(carboxymethyl)amino)acetic acid hydrate sodium
Synonyms: ({[3-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-2,5-dimethylphenyl)-2,2-dioxo-1,2λ⁶-benzoxathiol-3-yl]methyl}(carboxymethyl)amino)acetic acid hydrate sodium
Molecular Weight (g/mol): 621.57
Molecular Formula: C25H30N2NaO13S
InChi Key: NHDTXEGRWRPXMN-UHFFFAOYNA-N
SMILES: O.[Na].CC1=C(O)C(CN(CC(O)=O)CC(O)=O)=C(C)C(=C1)C1(CN(CC(O)=O)CC(O)=O)C2=CC=CC=C2OS1(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results