Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylxylenol Blue 85.0+%, TCI America™
SDP

Catalog No. X00521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X00521G Supplier TCI America Supplier No. X00521G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29412-85-9
Molecular Formula C25H29N2NaO13S
Molecular Weight (g/mol) 620.558
MDL Number MFCD00065250
InChI Key VYPGOJHOWVUMIQ-UHFFFAOYSA-M
PubChem CID 131674796
IUPAC Name sodium;2-[[5-[3-[[bis(carboxymethyl)amino]methyl]-2,2-dioxo-1,2$l^{6}-benzoxathiol-3-yl]-2-hydroxy-3,6-dimethylphenyl]methyl-(carboxymethyl)amino]acetate;hydrate
SMILES CC1=C(C(=C(C(=C1)C2(C3=CC=CC=C3OS2(=O)=O)CN(CC(=O)O)CC(=O)O)C)CN(CC(=O)O)CC(=O)[O-])O.O.[Na+]

Specifications

Quantity 1 g
Formula Weight 788.64
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methylxylenol Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.