accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SBr

CAS RN 29421-92-9

IUPAC Name: 4-bromo-2-methylthiophene
Synonyms: 4-bromo-2-methylthiophene
Molecular Weight (g/mol): 177.06
Molecular Formula: C5H5BrS
InChi Key: ABMUSXPGSSMPLK-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results