Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-methylthiophene 98.0+%, TCI America™
SDP

Catalog No. B33855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33855G 5 g
B338525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33855G Supplier TCI America Supplier No. B33855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29421-92-9
Molecular Formula C5H5BrS
Molecular Weight (g/mol) 177.059
MDL Number MFCD04116367
InChI Key ABMUSXPGSSMPLK-UHFFFAOYSA-N
PubChem CID 638584
IUPAC Name 4-bromo-2-methylthiophene
SMILES CC1=CC(=CS1)Br

Specifications

Color Yellow
Boiling Point 62°C
Quantity 5 g
Formula Weight 177.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-methylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.