accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29426-64-0

IUPAC Name: 3-aminoazepan-2-one hydrochloride
Synonyms: α-amino-epsilon-caprolactam hydrochloride
Molecular Weight (g/mol): 164.63
Molecular Formula: C6H13ClN2O
InChi Key: LWXJCGXAYXXXRU-UHFFFAOYNA-N
SMILES: Cl.NC1CCCCNC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results