Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-alpha-Amino-epsilon-caprolactam Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. A23315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23315G Supplier TCI America Supplier No. A23315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29426-64-0
Molecular Formula C6H13ClN2O
Molecular Weight (g/mol) 164.63
MDL Number MFCD11840204
InChI Key LWXJCGXAYXXXRU-UHFFFAOYNA-N
Synonym DL-3-Aminohexahydro-2-azepinone Hydrochloride, DL-3-Amino-2-oxohexamethyleneimine Hydrochloride
PubChem CID 12228562
IUPAC Name 3-aminoazepan-2-one hydrochloride
SMILES Cl.NC1CCCCNC1=O

Specifications

Melting Point 294°C
Color White-Yellow
Quantity 5 g
Formula Weight 164.63
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-alpha-Amino-epsilon-caprolactam Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.