accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOCH3

CAS RN 2946-61-4

IUPAC Name: dimethyl phenylphosphonite
Synonyms: dimethyl phenylphosphonite
Molecular Weight (g/mol): 170.15
Molecular Formula: C8H11O2P
InChi Key: LMZLQYYLELWCCW-UHFFFAOYSA-N
SMILES: COP(OC)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)