Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-(Boc-Amino)-1-Hexanol

Catalog No. 153045G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-304-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-304-5G Supplier Sigma Aldrich Supplier No. 153045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

6-(Boc-amino)-1-hexanol can be used as a reactant to synthesize:Hsp90 fluorescent probes applicable in the development of an FP assay for the Hsp90 chaperone.1-Guanidino-7-aminoheptane (GC7) analogs as potent deoxyhypusine synthase inhibitors.Linker; preparation of the iodide

Specifications

CAS 2946-61-4
Density 1.072 g/mL at 25°C
Molecular Formula C8H11O2P
Linear Formula C6H5P(OCH3)2
MDL Number MFCD00008352
Quantity 5 g
Molecular Weight (g/mol) 170.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.