CAS RN 294891-81-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Dvl-PDZ Domain Inhibitor II
The Dvl-PDZ Domain Inhibitor II, also referenced under CAS 294891-81-9, controls the biological activity of Dvl-PDZ. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.