Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Dvl-PDZ Domain Inhibitor II

Catalog No. 32233810MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-233-810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-233-810MG Supplier MilliporeSigma™ Supplier No. 32233810MG
Add to Cart
Add to Cart

The Dvl-PDZ Domain Inhibitor II, also referenced under CAS 294891-81-9, controls the biological activity of Dvl-PDZ. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
CAS 294891-81-9
Molecular Formula C22H18N2O4
Formula Weight 374.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.