accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2949-26-0

IUPAC Name: 2-ethynylnaphthalene
Synonyms: 2-ethynylnaphthalene
Molecular Weight (g/mol): 152.2
Molecular Formula: C12H8
InChi Key: IZXPFTLEVNQLGD-UHFFFAOYSA-N
SMILES: C#CC1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results