Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-ethynyl-naphthalene

Catalog No. CDS008002 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS008002 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS008002 Supplier Sigma Aldrich Supplier No. CDS008002250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Fluorobenzyl alcohol undergoes acetylation reaction with acetic anhydride catalyzed by tribromo melamine to yield 3-fluorobenzyl acetate.3-Fluorobenzyl alcohol was used as substrate to study pH dependence of aryl-alcohol oxidase activity.

Specifications

CAS 2949-26-0
Molecular Formula C12H8
Linear Formula C12H8
MDL Number MFCD00870424
Quantity 5 g
Molecular Weight (g/mol) 152.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.