accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSOOFFF

CAS RN 29540-83-8

IUPAC Name: 4-methylphenyl trifluoromethanesulfonate
Synonyms: 4-methylphenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 240.2
Molecular Formula: C8H7F3O3S
InChi Key: XXSBXVHIYGQWDV-UHFFFAOYSA-N
SMILES: CC1=CC=C(OS(=O)(=O)C(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results