Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolyl Trifluoromethanesulfonate 95.0+%, TCI America™
SDP

Catalog No. T26065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T26065G Supplier TCI America Supplier No. T26065G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29540-83-8
Molecular Formula C8H7F3O3S
Molecular Weight (g/mol) 240.196
MDL Number MFCD00192507
InChI Key XXSBXVHIYGQWDV-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid p-Tolyl Ester, p-Tolyl Triflate, 4-Methylphenyl Trifluoromethanesulfonate
PubChem CID 572481
IUPAC Name (4-methylphenyl) trifluoromethanesulfonate
SMILES CC1=CC=C(C=C1)OS(=O)(=O)C(F)(F)F

Specifications

Boiling Point 115°C
Quantity 5 g
Formula Weight 240.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Tolyl Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.