accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29559-52-2

IUPAC Name: 1,1,2-trichloro-2-methylpropane
Synonyms: 1,1,2-trichloro-2-methylpropane
Molecular Weight (g/mol): 161.45
Molecular Formula: C4H7Cl3
InChi Key: FRRHZKFKOHHEJR-UHFFFAOYSA-N
SMILES: CC(C)(Cl)C(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results