Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2-Trichloro-2-methylpropane, TCI America™
SDP

Catalog No. T116825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T116825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T116825G Supplier TCI America Supplier No. T116825G

Chemical Identifiers

CAS 29559-52-2
Molecular Formula C4H7Cl3
Molecular Weight (g/mol) 161.45
MDL Number MFCD00059916
InChI Key FRRHZKFKOHHEJR-UHFFFAOYSA-N
PubChem CID 520441
IUPAC Name 1,1,2-trichloro-2-methylpropane
SMILES CC(C)(C(Cl)Cl)Cl

Specifications

Color Yellow
Boiling Point 162°C
Quantity 25 g
Formula Weight 161.45
Physical Form Liquid
Chemical Name or Material 1,1,2-Trichloro-2-methylpropane
EINECSNumber (2)-0062
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.