accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2959-96-8

IUPAC Name: 3-phenyloxane-2,6-dione
Synonyms: 3-phenyloxane-2,6-dione
Molecular Weight (g/mol): 190.2
Molecular Formula: C11H10O3
InChi Key: NVPRNSAYSSEIGR-UHFFFAOYNA-N
SMILES: O=C1CCC(C(=O)O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results