Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylglutaric Anhydride 97%, Thermo Scientific™

Catalog No. AC130620010
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC130620010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC130620010 Supplier Thermo Scientific Chemicals Supplier No. 130620010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2959-96-8
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C11H10O3
Quantity 1g
MDL Number MFCD00006681
Beilstein 17, 494
Synonym 2-phenylglutaric anhydride, 3-phenyldihydro-2h-pyran-2,6 3h-dione, 2h-pyran-2,6 3h-dione, dihydro-3-phenyl, .alpha.-phenylglutaric anhydride, 2-phenylglutaric anhydride treated bsa, 2-phenylglutaricanhydride, 3-phenyl-dihydro-pyran-2,6-dione, dihydro-3-phenyl-2h-pyran-2,6 3h dione, 2h-pyran-2,6 3h-dione,dihydro-3-phenyl
InChI Key NVPRNSAYSSEIGR-UHFFFAOYSA-N
SMILES C1CC(=O)OC(=O)C1C2=CC=CC=C2
IUPAC Name 3-phenyloxane-2,6-dione
Molecular Weight (g/mol) 190.198
PubChem CID 96293
Formula Weight 190.2
Percent Purity ≥96.0%
Chemical Name or Material 2-Phenylglutaric anhydride, 97%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 220-991-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.