CAS RN 296-35-5
IUPAC Name:
1,4,7,10,13,16-hexaazacyclooctadecane
Synonyms:
1,4,7,10,13,16-hexaazacyclooctadecane
Molecular Weight (g/mol):
258.41
Molecular Formula:
C12H30N6
InChi Key:
RVJABZUDCPZPPM-UHFFFAOYSA-N
SMILES:
C1CNCCNCCNCCNCCNCCN1
1
–
1
of
1
results
1
Sigma Aldrich 3-(Benzyloxy)Phenylboronic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.