accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29604-34-0

IUPAC Name: zinc(2+) dichloride
Synonyms: ZnCl2|zinc chloride, anhydrous|zinc dichloride|butter of zinc|zinkchlorid|chlorure de zinc|dichlorozinc
Molecular Weight (g/mol): 136.28
Molecular Formula: Cl2Zn
InChi Key: JIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Zn++]
Physical Form 
  • (6)
Quantity 
  • (2)
  • (2)
  • (2)
Grade 
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)