CAS RN 2961-50-4
IUPAC Name:
4-pentylpyridine
Synonyms:
P-amylpyridine
Molecular Weight (g/mol):
149.24
Molecular Formula:
C10H15N
InChi Key:
ABJVUPUJUGBUMM-UHFFFAOYSA-N
SMILES:
CCCCCC1=CC=NC=C1
1
–
1
of
1
results
1
4-Amylpyridine 98.0+%, TCI America™
CAS: 2961-50-4 Molecular Formula: C10H15N Molecular Weight (g/mol): 149.237 MDL Number: MFCD02093422 InChI Key: ABJVUPUJUGBUMM-UHFFFAOYSA-N Synonym: 4-Pentylpyridine PubChem CID: 72918 IUPAC Name: 4-pentylpyridine SMILES: CCCCCC1=CC=NC=C1