Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amylpyridine 98.0+%, TCI America™
SDP

Catalog No. A14985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14985G Supplier TCI America Supplier No. A14985G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2961-50-4
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD02093422
InChI Key ABJVUPUJUGBUMM-UHFFFAOYSA-N
Synonym 4-Pentylpyridine
PubChem CID 72918
IUPAC Name 4-pentylpyridine
SMILES CCCCCC1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 230°C
Quantity 5 g
Formula Weight 149.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Amylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.