accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29621-88-3

H2N(R)SeSe(R)NH2HOOOHO

CAS RN 29621-88-3

IUPAC Name: (2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]diselanyl}propanoic acid
Synonyms: (2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]diselanyl}propanoic acid
Molecular Weight (g/mol): 334.11
Molecular Formula: C6H12N2O4Se2
InChi Key: JULROCUWKLNBSN-IMJSIDKUSA-N
SMILES: N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)