Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Selenocystine Monohydrate, TCI America™
SDP

Catalog No. S0900250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0900250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0900250MG Supplier TCI America Supplier No. S0900250MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29621-88-3
Molecular Formula C6H12N2O4Se2
Molecular Weight (g/mol) 334.11
MDL Number MFCD00800971
InChI Key JULROCUWKLNBSN-IMJSIDKUSA-N
Synonym l-selenocystine, seleno-l-cystine, l-3,3'-diselenodialanine, alanine, 3,3'-diselenodi-, l, selenocystine, l, l-alanine,3,3'-diselenobis, 2r-2-amino-3-2r-2-amino-2-carboxyethyl diselanyl propanoic acid, l-selenocystine monohydrate, selenocystine, l-isomer
PubChem CID 207306
IUPAC Name (2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]diselanyl}propanoic acid
SMILES N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O

Specifications

Melting Point 219°C
UN Number 3283
Quantity 250 mg
Formula Weight 334.09
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Selenocystine Monohydrate
RTECSNumber AY6032000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.